missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-23R Antibody (15N6C6), DyLight 755, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-27092IR
This item is not returnable.
View return policy
Description
IL-23R Monoclonal specifically detects IL-23R in Human, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| IL-23R | |
| Monoclonal | |
| DyLight 755 | |
| Q5VWk5 | |
| IL23R | |
| A synthetic peptide corresponding to amino acids 288-315 of hIL-23 receptor was used as immunogen for this antibody. | |
| 0.1 mL | |
| Immunology | |
| 149233 | |
| Human, Primate | |
| Purified |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 15N6C6 | |
| Western Blot, Immunohistochemistry-Paraffin | |
| IBD17, IL-23 receptor, IL-23R, interleukin 23 receptor, interleukin-23 receptor | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| There are multiple isoforms of IL-23 receptor. Researchers are encouraged to use a tool like BLAST to determine the suitability of this antibody for their testing. | |
| Store at 4C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction