missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-23R Antibody (218213) [HRP], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB14001H
This item is not returnable.
View return policy
Description
IL-23R Monoclonal antibody specifically detects IL-23R in Human samples. It is validated for Western Blot
Specifications
| IL-23R | |
| Monoclonal | |
| HRP | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| 218213 | |
| Western Blot | |
| IBD17, IL-23 receptor, IL-23R, interleukin 23 receptor, interleukin-23 receptor | |
| Mouse myeloma cell line NS0-derived recombinant human IL-23 R, Gly24-Ile354, Accession # Q5VWK5 | |
| 0.1 mL | |
| Immunology | |
| 149233 | |
| Store at 4°C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction