Learn More
IL-24 Antibody (101), mFluor Violet 450 SE, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90083MFV450
This item is not returnable.
View return policy
Description
IL-24 Monoclonal antibody specifically detects IL-24 in Human samples. It is validated for ELISA
Specifications
| IL-24 | |
| mFluor Violet 450 SE | |
| 0.1 mL |
| Monoclonal | |
| Protein A purified | |
| Store at 4C in the dark. |