missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-24 Antibody (MM0398-6F48), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-11702AF532
This item is not returnable.
View return policy
Description
IL-24 Monoclonal specifically detects IL-24 in Human samples. It is validated for Western Blot.
Specifications
| IL-24 | |
| Monoclonal | |
| Alexa Fluor 532 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| IL24 | |
| Human recombinant IL-24 | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG2 |
| Western Blot | |
| MM0398-6F48 | |
| Western Blot | |
| C49A, FISP, IL10B, IL-24, interleukin 24, mda-7, MDA7MOB5, melanocyte-associated Mda-7, melanoma differentiation association protein 7, Melanoma differentiation-associated gene 7 protein, Mob-5, ST16interleukin-24, suppression of tumorigenicity 16 (melanoma differentiation), Suppression of tumorigenicity 16 protein | |
| Mouse | |
| Protein G purified | |
| Apoptosis, Cancer | |
| 11009.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction