missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IL-25 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5100835
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total IL25.
Interleukin 25 (IL-25) is a member of the IL-17 family and is identified as a proinflammatory cytokine which is produced by Th17 effector cells thus enabling the Th17 subset to initiate or maintain the autoimmune process in diseases such as Rheumatoid Arthritis. IL-25 also induces activation of NF-kappa-B and stimulates production of the proinflammatory chemokine IL-8. IL-25 itself is activated through IL-23, IL-5, and TGF-beta.
Specifications
| IL-25 | |
| Polyclonal | |
| Unconjugated | |
| IL25 | |
| D17Wsu104e; il 17e; Il17e; IL-17E; Il25; IL-25; ILN; Interleukin; interleukin 17E; interleukin 25; interleukin-17E; Interleukin-25; Ly6elg; Mydgf; myeloid derived growth factor; myeloid-derived growth factor; RGD1561632; UNQ3120/PRO10272; UPF0556 protein C19orf10 homolog | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 140806, 64806 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9H293 | |
| IL25 | |
| A synthesized peptide derived from human IL25(Accession Q9H293), corresponding to amino acid residues G44-L94. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction