missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-27 Antibody (307426), Alexa Fluor™ 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB25261G
This item is not returnable.
View return policy
Description
IL-27 Monoclonal antibody specifically detects IL-27 in Human samples. It is validated for Flow Cytometry, CyTOF
Specifications
| IL-27 | |
| Monoclonal | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, CyTOF | |
| 307426 | |
| Intracellular Staining by Flow Cytometry, CyTOF-ready | |
| IL-27, IL-27 p28 subunit, IL-27 subunit alpha, IL27A, IL-27-A, IL27-A, IL27p28, IL30, interleukin 27, interleukin 30, interleukin-27 subunit alpha, MGC71873, p28IL-27A | |
| Mouse myeloma cell line NS0-derived recombinant human IL-27, Arg21-Lys229 of EBI-3 (Accession #Q14213.2) and Phe29-Pro243 of p28 (Accession #AAM34498) | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 246778 | |
| Store at 4C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction