missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-27 Antibody, DyLight 488, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76698G
This item is not returnable.
View return policy
Description
IL-27 Polyclonal antibody specifically detects IL-27 in Human, Mouse samples. It is validated for Western Blot, ELISA
Specifications
| IL-27 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, ELISA | |
| DyLight 488 | |
| IL-27, IL-27 p28 subunit, IL-27 subunit alpha, IL27A, IL-27-A, IL27-A, IL27p28, IL30, interleukin 27, interleukin 30, interleukin-27 subunit alpha, MGC71873, p28IL-27A | |
| Antibody was raised against a peptide corresponding to 16 amino acids near the amino terminus of human IL-27. The immunogen is located within the first 50 amino acids of IL-27. Amino Acid Squence: WGFPRPPGRPQLSLQE | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 246778 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction