missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-27 p28 Polyclonal Antibody

IL-27 Polyclonal Antibody
Brand: Invitrogen PA578661
This item is not returnable.
View return policy
Description
IL-27 Polyclonal Antibody for Western Blot, IHC (P)
Specifications
| IL-27 p28 | |
| Polyclonal | |
| Unconjugated | |
| Il27 | |
| IL27; IL-27; IL-27 p28 subunit; IL-27 subunit alpha; Il27a; IL-27A; IL-27-A; IL27-A; IL27p28; IL-27p28; IL30; IL-30; ILN; Interleukin; interleukin 27; interleukin 30; Interleukin-27; interleukin-27 subunit alpha; Interleukin-30; p28 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 246778 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1.65 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q8NEV9 | |
| Il27 | |
| Recombinant protein encompassing a sequence within the center region of human IL27. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction