missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-27R alpha/WSX-1/TCCR Antibody (191106), DyLight 550, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB1479L
This item is not returnable.
View return policy
Description
IL-27R alpha/WSX-1/TCCR Monoclonal antibody specifically detects IL-27R alpha/WSX-1/TCCR in Human samples. It is validated for Western Blot
Specifications
| IL-27R alpha/WSX-1/TCCR | |
| Monoclonal | |
| DyLight 550 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| 191106 | |
| Western Blot | |
| class I cytokine receptor, CRL1IL-27R, Cytokine receptor-like 1, IL27R, IL-27RA, IL-27R-alpha, interleukin 27 receptor, alpha, interleukin-27 receptor subunit alpha, TCCRIL-27 receptor subunit alpha, T-cell cytokine receptor type 1, Type I T-cell cytokine receptor, WSX-1, WSX1IL-27R subunit alpha, zcytor1 | |
| Mouse myeloma cell line NS0-derived recombinant human IL-27 R alpha/WSX-1/TCCR, Gly34-Lys516, Accession # Q6UWB1 | |
| 0.5 mL | |
| Apoptosis, Cytokine Research | |
| 9466 | |
| Store at 4C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction