missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-27R alpha/WSX-1/TCCR Antibody (MM0563-8A5), Alexa Fluor™ 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-11917AF594
This item is not returnable.
View return policy
Description
IL-27R alpha/WSX-1/TCCR Monoclonal specifically detects IL-27R alpha/WSX-1/TCCR in Human samples. It is validated for Western Blot.
Specifications
| IL-27R alpha/WSX-1/TCCR | |
| Monoclonal | |
| Alexa Fluor 594 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| IL27RA | |
| Human recombinant IL-27R alpha/WSX-1/TCCR (extracellular domain) | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG2 |
| Western Blot | |
| MM0563-8A5 | |
| Western Blot | |
| class I cytokine receptor, CRL1IL-27R, Cytokine receptor-like 1, IL27R, IL-27RA, IL-27R-alpha, interleukin 27 receptor, alpha, interleukin-27 receptor subunit alpha, TCCRIL-27 receptor subunit alpha, T-cell cytokine receptor type 1, Type I T-cell cytokine receptor, WSX-1, WSX1IL-27R subunit alpha, zcytor1 | |
| Mouse | |
| Protein G purified | |
| Apoptosis, Cytokine Research | |
| 9466.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction