Learn More
IL-29/IFN-lambda 1 Antibody (B-Z29), mFluor Violet 450 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-18107MFV450
Product Code. 18542265
  • € 439.00 / 0.10mL
Description
IL-29/IFN-lambda 1 Monoclonal antibody specifically detects IL-29/IFN-lambda 1 in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ELISA
cytokine Zcyto21, IFNL1, IFN-lambda-1, IL-29, interferon lambda-1, interferon, lambda 1, interleukin 29 (interferon, lambda 1), interleukin-29, ZCYTO21
Primary
Purified
mFluor Violet 450 SE
Mouse
282618
IgG2a κ