missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-29/IFN-lambda 1 Antibody, DyLight 755, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-77893IR
This item is not returnable.
View return policy
Description
IL-29/IFN-lambda 1 Polyclonal antibody specifically detects IL-29/IFN-lambda 1 in Human samples. It is validated for Western Blot, ELISA
Specifications
| IL-29/IFN-lambda 1 | |
| Polyclonal | |
| Western Blot, ELISA | |
| cytokine Zcyto21, IFNL1, IFN-lambda-1, IL-29, interferon lambda-1, interferon, lambda 1, interleukin 29 (interferon, lambda 1), interleukin-29, ZCYTO21 | |
| This IgG fraction IL-29/IFN-lambda 1 Antibody was prepared from rabbit antiserum after repeated immunizations with mature length recombinant human IL-29/IFN-lambda 1/IFN-lambda 1 protein produced in E.coli. (Uniprot: Q8IU54) | |
| 0.1 mL | |
| Biologically Active Proteins | |
| 282618 | |
| Store at 4C in the dark. |
| Western Blot, ELISA | |
| DyLight 755 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Multi-step | |
| RUO | |
| Primary | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction