Learn More
IL-31 Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP1-76419CL1
This item is not returnable.
View return policy
Description
IL-31 Polyclonal antibody specifically detects IL-31 in Human samples. It is validated for ELISA,Western Blot,Immunocytochemistry/Immunofluorescence
Specifications
| IL-31 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA, Western Blot, Immunocytochemistry/Immunofluorescence | |
| CoraFluor 1 | |
| IL-31interleukin-31, interleukin 31 | |
| Antibody was raised against a 18 amino acid synthetic peptide from near the center of human IL-31. The immunogen is located within amino acids 60 - 110 of IL-31. Amino Acid Squence: CLSPDAQPPNNIHSPAIR | |
| 0.1 mL | |
| Biologically Active Proteins | |
| 386653 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |