missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-32 Antibody (034), mFluor Violet 450 SE, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89776MFV450
This item is not returnable.
View return policy
Description
IL-32 Monoclonal antibody specifically detects IL-32 in Human samples. It is validated for ELISA, ELISA
Specifications
| IL-32 | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA, Sandwich ELISA | |
| 034 | |
| ELISA, Sandwich ELISA Capture | |
| IL-32, IL-32alpha, IL-32beta, interleukin 32, interleukin-32, interleukin-32 theta, natural killer cell transcript 4, Natural killer cells protein 4, NK4IL-32delta, TAIFa, TAIFb, TAIFc, TAIFd, TAIFIL-32gamma, Tumor necrosis factor alpha-inducing factor | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human IL-32 (Accession#: NP_001012651.1; Met1-Lys131). | |
| 0.1 mL | |
| Cancer, Signal Transduction | |
| 9235 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction