missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-32 Antibody (373821), PE, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB30402P
This item is not returnable.
View return policy
Description
IL-32 Monoclonal antibody specifically detects IL-32 in Human samples. It is validated for Flow Cytometry (Intracellular Staining), CyTOF
Specifications
| IL-32 | |
| Monoclonal | |
| PE | |
| PBS | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry (Intracellular Staining), CyTOF | |
| 373821 | |
| Intracellular Staining by Flow Cytometry, CyTOF-ready | |
| IL-32, IL-32alpha, IL-32beta, interleukin 32, interleukin-32, interleukin-32 theta, natural killer cell transcript 4, Natural killer cells protein 4, NK4IL-32delta, TAIFa, TAIFb, TAIFc, TAIFd, TAIFIL-32gamma, Tumor necrosis factor alpha-inducing factor | |
| E. coli-derived recombinant human IL-32 alpha, Met1-Asn131, Accession # P24001 | |
| 0.1 mL | |
| Cancer, Signal Transduction | |
| 9235 | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction