missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-32 Antibody, PE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76684PE
This item is not returnable.
View return policy
Description
IL-32 Polyclonal antibody specifically detects IL-32 in Human samples. It is validated for Flow Cytometry
Specifications
| IL-32 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| PE | |
| IL-32, IL-32alpha, IL-32beta, interleukin 32, interleukin-32, interleukin-32 theta, natural killer cell transcript 4, Natural killer cells protein 4, NK4IL-32delta, TAIFa, TAIFb, TAIFc, TAIFd, TAIFIL-32gamma, Tumor necrosis factor alpha-inducing factor | |
| Antibody was raised against a peptide corresponding to 15 amino acids near the carboxy terminus of human IL-32. The immunogen is located within the last 50 amino acids of IL-32. Amino Acid Sequence: YGAPRGDKEELTPQK | |
| 0.1 mL | |
| Cancer, Signal Transduction | |
| 9235 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction