Learn More
IL-33 Antibody (396118), mFluor Violet 610 SE, Novus Biologicals™
Rat Monoclonal Antibody
Brand:  Novus Biologicals FAB3626MFV610
Product Code. 18691560
  • € 719.00 / 0.10mL
Define Immunology Research on Your Terms With R&D Systems
Use promo code 24090
View all promotions
Description
IL-33 Monoclonal antibody specifically detects IL-33 in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IL-33
Monoclonal
mFluor Violet 610 SE
50mM Sodium Borate
Rat
0.1 mL
Biologically Active Proteins, Cellular Markers, Core ESC Like Genes, Immunology, Inflammation, Innate Immunity, Neuroscience, Sensory Systems, Stem Cell Markers, Stem Cells, Transcription Factors and Regulators, Vision
90865
Store at 4C in the dark.
Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF
396118
Western Blot, Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready
C9orf26chromosome 9 open reading frame 26 (NF-HEV), DKFZp586H0523, DVS27, DVS27-related protein, IL1F11, IL-1F11, IL-33, interleukin 33, Interleukin-1 family member 11, interleukin-33, NFHEV, NF-HEVNFEHEV, Nuclear factor from high endothelial venules, RP11-575C20.2
E. coli-derived recombinant mouse IL-33, Ser109-Ile266, Accession # Q8BVZ5