missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
IL-33 Monoclonal antibody specifically detects IL-33 in Human samples. It is validated for Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence,Immunohistochemistry
Specifications
Specifications
| Antigen | IL-33 |
| Applications | Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence, Immunohistochemistry |
| Classification | Monoclonal |
| Clone | CL13438 |
| Concentration | LYOPH |
| Conjugate | Unconjugated |
| Dilution | Immunocytochemistry/ Immunofluorescence 2-10 μg/mL, Immunohistochemistry-Paraffin 1:500 - 1:1000 |
| Formulation | Lyophilized in 5% Trehalose. |
| Gene Alias | C9orf26chromosome 9 open reading frame 26 (NF-HEV), DKFZp586H0523, DVS27, DVS27-related protein, IL1F11, IL-1F11, IL-33, interleukin 33, Interleukin-1 family member 11, interleukin-33, NFHEV, NF-HEVNFEHEV, Nuclear factor from high endothelial venules, RP11-575C20.2 |
| Host Species | Mouse |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?