Learn More
IL-34 Antibody (1D12), Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP1-51627
Product Code. 18745623
  • € 498.00 / 0.10mL
Description
IL-34 Monoclonal specifically detects IL-34 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Monoclonal
Unpurified
IgG1
IL34
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.