missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-34 Antibody (36W18), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-21982H
This item is not returnable.
View return policy
Description
IL-34 Monoclonal specifically detects IL-34 in Human samples. It is validated for ELISA.
Specifications
| IL-34 | |
| Monoclonal | |
| HRP | |
| PBS with No Preservative | |
| IL34 | |
| Human recombinant IL-34 | |
| 0.1 mL | |
| 146433 | |
| Store at 4C in the dark. | |
| IgG1 |
| ELISA | |
| 36W18 | |
| ELISA | |
| C16orf77, IL-34, interleukin 34 | |
| Mouse | |
| Protein G purified | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction