missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-34 Antibody, mFluor Violet 610 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76357MFV610
This item is not returnable.
View return policy
Description
IL-34 Polyclonal antibody specifically detects IL-34 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA
Specifications
| IL-34 | |
| Polyclonal | |
| Western Blot, ELISA | |
| C16orf77, IL-34, interleukin 34 | |
| Antibody was raised against a 16 amino acid synthetic peptide from near the amino terminus of human IL-34. The immunogen is located within amino acids 30 - 80 of IL-34. Amino Acid Squence: LRDKLQYRSRLQYMKH | |
| 0.1 mL | |
| Signal Transduction | |
| 146433 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, ELISA | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction