missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-36 alpha/IL-1F6 Antibody, HRP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-41238H
This item is not returnable.
View return policy
Description
IL-36 alpha/IL-1F6 Polyclonal antibody specifically detects IL-36 alpha/IL-1F6 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| IL-36 alpha/IL-1F6 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| HRP | |
| FIL1, FIL1 epsilon, FIL1(EPSILON), FIL1E, IL-1 epsilon, IL1(EPSILON), IL1E, IL1F6, IL-1F6, IL-1F6 (FIL-1-epsilon), interleukin 1 family, member 6 (epsilon), interleukin 1, epsilon, Interleukin 36, Alpha, Interleukin-1 epsilon, interleukin-1 family member 6, Interleukin-36 Alpha | |
| Antibody was raised against an 11 amino acid peptide near the carboxy terminus of human IL-36A. The immunogen is located within the last 50 amino acids of IL-36A. Amino Acid Squence: AELGKANTTDFG | |
| 0.1 mL | |
| Biologically Active Proteins | |
| 27179 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction