Learn More
IL-36 Beta/IL-1F8 Mouse anti-Human, CoraLite 488, Clone: 10D12B1, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-60290-100UL
Description
The IL-36 Beta/IL-1F8 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human IL-36 Beta/IL-1F8. This antibody recognizes human antigen. The IL-36 Beta/IL-1F8 antibody has been validated for the following applications: IF analysis. This antibody is coralite™488.
Specifications
| IL-36 Beta/IL-1F8 | |
| Monoclonal | |
| CoraLite 488 | |
| 100 μL | |
| 27177 | |
| IgG1 |
| Immunofluorescence | |
| 10D12B1 | |
| Mouse | |
| Primary | |
| Human |