Learn More
IL-36 Beta/IL-1F8 Mouse anti-Human, CoraLite 488, Clone: 10D12B1, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech CL488-60290-100UL
Product Code. 17462543
  • € 450.00 / 100µL
Description
The IL-36 Beta/IL-1F8 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human IL-36 Beta/IL-1F8. This antibody recognizes human antigen. The IL-36 Beta/IL-1F8 antibody has been validated for the following applications: IF analysis. This antibody is coralite™488.
Specifications
Specifications
IL-36 Beta/IL-1F8
Monoclonal
CoraLite 488
100 μL
27177
IgG1
Immunofluorescence
10D12B1