missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-36Ra/IL-1F5 Antibody, HRP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98241H
This item is not returnable.
View return policy
Description
IL-36Ra/IL-1F5 Polyclonal specifically detects IL-36Ra/IL-1F5 in Mouse samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| IL-36Ra/IL-1F5 | |
| Polyclonal | |
| ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| FIL1, FIL1 delta, FIL1-delta, IL-1RP3, IL-1L1, IL-1-delta), FIL1DIL-1 delta, IL1F5 (Canonical product IL-1F5a), IL-1RP3, IL36RA, IL-36RA, interleukin 1 family, member 5 (delta), interleukin 1, delta, Interleukin 36 Receptor Antagonist, interleukin-1 family member 5, Interleukin-1 HY1, Interleukin-1 receptor antagonist homolog 1, Interleukin-1-like protein 1, Interleukin-36 Receptor Antagonist | |
| Produced in rabbits immunized with purified, recombinant IL-36Ra/IL-1F5 (NP_062324.2; Val3-Asp156). | |
| 0.1 mL | |
| Biologically Active Proteins | |
| 26525 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| HRP | |
| PBS | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction