missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-37/IL-1F7 Antibody (899826), Alexa Fluor™ 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB19751G
This item is not returnable.
View return policy
Description
IL-37/IL-1F7 Monoclonal antibody specifically detects IL-37/IL-1F7 in Human samples. It is validated for Flow Cytometry, CyTOF
Specifications
| IL-37/IL-1F7 | |
| Monoclonal | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, CyTOF | |
| 899826 | |
| Flow Cytometry, CyTOF-ready | |
| FIL1, FIL1(ZETA), FIL1ZFIL1 zeta, IL-1F7IL-1 zeta, IL1H4IL1F7 (canonical product IL-1F7b), IL-1H4IL-1F7b (IL-1H4, IL-1H, IL-1RP1), IL-1RP1IL-1X protein, IL1RP1interleukin 1 family member 7, IL-1X, interleukin 1 family, member 7 (zeta), interleukin 1, zeta, interleukin-1 family member 7, Interleukin-1 homolog 4, interleukin-1 superfamily z, Interleukin-1 zeta, Interleukin-1-related protein | |
| E. coli-derived recombinant human IL-37/IL-1F7, Lys53-Val206, Accession # Q9NZH6 | |
| 0.1 mL | |
| Cytokine Research | |
| 27178 | |
| Store at 4C in the dark. | |
| IgG2b |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu