missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-37/IL-1F7 Antibody (B-N37), DyLight 755, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-18119IR
This item is not returnable.
View return policy
Description
IL-37/IL-1F7 Monoclonal antibody specifically detects IL-37/IL-1F7 in Human samples. It is validated for ELISA
Specifications
| IL-37/IL-1F7 | |
| Monoclonal | |
| DyLight 755 | |
| FIL1, FIL1(ZETA), FIL1ZFIL1 zeta, IL-1F7IL-1 zeta, IL1H4IL1F7 (canonical product IL-1F7b), IL-1H4IL-1F7b (IL-1H4, IL-1H, IL-1RP1), IL-1RP1IL-1X protein, IL1RP1interleukin 1 family member 7, IL-1X, interleukin 1 family, member 7 (zeta), interleukin 1, zeta, interleukin-1 family member 7, Interleukin-1 homolog 4, interleukin-1 superfamily z, Interleukin-1 zeta, Interleukin-1-related protein | |
| Recombinant human IL-37/IL-1F7. Hybridoma: Myeloma X63/AG.8653 x Balb/c node cells | |
| 0.1 mL | |
| Cytokine Research | |
| 27178 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| ELISA | |
| B-N37 | |
| 50mM Sodium Borate | |
| Mouse | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction