missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-38/IL-1F10 Antibody (798036R) [Janelia Fluor™ 585], Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB7774RJF585
This item is not returnable.
View return policy
Description
IL-38/IL-1F10 Monoclonal antibody specifically detects IL-38/IL-1F10 in Mouse samples. It is validated for Flow Cytometry,ELISA,Western Blot
Specifications
| IL-38/IL-1F10 | |
| Monoclonal | |
| Janelia Fluor 585 | |
| interleukin 1 family, member 10 (theta), UNQ6119/PRO20041 | |
| E.coli-derived recombinant mouse IL-38/IL-1F10, Met1-Arg152, Accession # Q8R459 | |
| RUO | |
| Primary | |
| Mouse | |
| Liquid |
| Flow Cytometry, ELISA, Western Blot | |
| 798036R | |
| 50mM Sodium Borate | |
| Rat | |
| 0.1 mL | |
| Immunology | |
| 84639 | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction