missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-3R beta Antibody (130705), Biotin, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB5492B
This item is not returnable.
View return policy
Description
IL-3R beta Monoclonal antibody specifically detects IL-3R beta in Mouse samples. It is validated for Flow Cytometry, CyTOF
Specifications
| IL-3R beta | |
| Monoclonal | |
| Biotin | |
| PBS | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 1439 | |
| Store at 4C in the dark. | |
| IgG2a |
| Flow Cytometry, CyTOF | |
| 130705 | |
| Flow Cytometry, CyTOF-ready | |
| AIC2A, BetaIl3, Bil3, Csfgmrb, IL3R, IL3RB, Il3rb2 | |
| Mouse myeloma cell line NS0-derived recombinant mouse IL-3 R beta , His23-Trp440, Accession # P26954 | |
| 0.5 mL | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction