missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-4 Antibody (99605) [Janelia Fluor™ 585], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB6543JF585
This item is not returnable.
View return policy
Description
IL-4 Monoclonal antibody specifically detects IL-4 in Porcine samples. It is validated for Western Blot,Flow Cytometry (Intracellular Staining),CyTOF
Specifications
| IL-4 | |
| Monoclonal | |
| Janelia Fluor 585 | |
| B cell growth factor 1, B-cell stimulatory factor 1, BCGF1, BCGF-1, binetrakin, BSF1, BSF-1, IL-4B_cell stimulatory factor 1, interleukin 4, interleukin-4, Lymphocyte stimulatory factor 1, MGC79402, Pitrakinra | |
| E. coli-derived recombinant porcine IL-4, His25-Cys133, Accession # Q04745 | |
| RUO | |
| Primary | |
| Pig | |
| Liquid |
| Western Blot, Flow Cytometry (Intracellular Staining), CyTOF | |
| 99605 | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Adaptive Immunity, Asthma, Biologically Active Proteins, Cancer, Cytokine Research, Immunology, Innate Immunity | |
| 3565 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction