missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-4I1 Antibody (1006231), Alexa Fluor™ 488, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB56843G
This item is not returnable.
View return policy
Description
IL-4I1 Monoclonal antibody specifically detects IL-4I1 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF
Specifications
| IL-4I1 | |
| Monoclonal | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, CyTOF | |
| 1006231 | |
| Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready | |
| FIG1, Fig-1 protein, hFIG1, IL4-induced protein 1, interleukin 4 induced 1, interleukin four induced 1, LAAO, L-amino-acid oxidase, LAO, UNQ636/PRO1265 | |
| Chinese Hamster Ovary cell line, CHO-derived human IL-4I1, Met1-His567, Accession # Q96RQ9 | |
| 0.1 mL | |
| Apoptosis, Cell Biology | |
| 259307 | |
| Store at 4C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction