Learn More
IL-4I1 Antibody (1006231) [DyLight 488], Novus Biologicals™
Rat Monoclonal Antibody
Brand:  Novus Biologicals FAB56843K
Product Code. 18529134
  • € 593.00 / 0.10mL
Description
IL-4I1 Monoclonal antibody specifically detects IL-4I1 in Human samples. It is validated for Western Blot, Flow Cytometry (Intracellular Staining), CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IL-4I1
Monoclonal
DyLight 488
50mM Sodium Borate
Rat
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
Human
Purified
Western Blot, Flow Cytometry (Intracellular Staining), CyTOF
1006231
Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready
FIG1, Fig-1 protein, hFIG1, IL4-induced protein 1, interleukin 4 induced 1, interleukin four induced 1, LAAO, L-amino-acid oxidase, LAO, UNQ636/PRO1265
Chinese Hamster Ovary cell line, CHO-derived human IL-4I1, Met1-His567, Accession # Q96RQ9
0.1 mL
Apoptosis, Cell Biology
259307
Store at 4°C in the dark.
IgG2a