Learn More
IL-4R alpha Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP3-21332-25ul
 View more versions of this product
Product Code. 18682614
  • € 401.00 / 25µL
Description
IL-4R alpha Polyclonal antibody specifically detects IL-4R alpha in Human samples. It is validated for Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IL-4R alpha
Polyclonal
Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml
CD124 antigen, IL-4 receptor subunit alpha, IL-4R subunit alpha, IL-4RA, IL4RACD124, IL-4R-alpha, interleukin 4 receptor, interleukin-4 receptor alpha chain, interleukin-4 receptor subunit alpha
This antibody was developed against Recombinant Protein corresponding to amino acids: DNLTCTETPLVIAGNPAYRSFSNSLSQSPCPRELGPDPLLARHLEEVEPEMPCVPQLSEPTTVPQPEPETWEQILRRNVLQHGAAAAPVSAPTSGYQEFV
25 μg
Asthma, B Cell Development and Differentiation Markers, Cancer, Immunology, Myeloid derived Suppressor Cell
3566
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
IgG
Immunofluorescence
Unconjugated
PBS, pH 7.2, 40% glycerol