missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-5 Antibody (mepolizumab), mFluor Violet 500 SE, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals NBP3-28010MFV500
This item is not returnable.
View return policy
Description
IL-5 Monoclonal antibody specifically detects IL-5 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| IL-5 | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| B-cell differentiation factor I, EDF, Eosinophil differentiation factor, IL-5T-cell replacing factor, interleukin 5 (colony-stimulating factor, eosinophil), interleukin-5, TRFB cell differentiation factor I | |
| IL-5 | |
| 0.1 mL | |
| Asthma, Biologically Active Proteins, Cell Cycle and Replication, Cytokine Research, Immunology | |
| 3567 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| mepolizumab | |
| 50mM Sodium Borate | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction