missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ CGRP1 Antibody (fremanezumab) - Humanized, IgG2SA, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-28123-100ug
This item is not returnable.
View return policy
Description
CGRP1 Monoclonal antibody specifically detects CGRP1 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| CGRP1 | |
| Monoclonal | |
| Unconjugated | |
| P06881 | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. | |
| IgG2 |
| Flow Cytometry, ELISA, Functional Assay | |
| fremanezumab | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| alpha-type CGRP, CALCA, calcitonin gene-related peptide I, calcitonin/calcitonin-related polypeptide, alpha, calcitonin; calcitonin gene-related peptide 1, CGRP-I | |
| CALCA/ CGRP | |
| 100 μg | |
| Cardiovascular Biology, Cellular Signaling, Neuroscience, Signal Transduction | |
| 796 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction