Learn More
IL-6 Antibody (Chugai SK2), CoraFluor™ 1, Novus Biologicals™
Human Monoclonal Antibody
Brand:  Novus Biologicals NBP3-28422CL1
Product Code. 30155751
  • € 869.00 / 0.10mL
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Description
Description
IL-6 Monoclonal antibody specifically detects IL-6 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY
Spécification
Spécification
IL-6
Monoclonal
CoraFluor 1
B-cell differentiation factor, B-cell stimulatory factor 2, BSF2, BSF-2, CDF, CTL differentiation factor, HSF, hybridoma growth factor, IFNB2, IFN-beta-2, IL-6, Interferon beta-2, interleukin 6 (interferon, beta 2), interleukin BSF-2, interleukin-6
IL-6/ IFNb2
0.1 mL
Adaptive Immunity, Biologically Active Proteins, Cancer, Cell Cycle and Replication, Cytokine Research, Immunology, Innate Immunity, Neuroscience
3569
Store at 4°C in the dark. Do not freeze.
IgG1
Flow Cytometry, ELISA, Functional Assay
Chugai SK2