missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-6 Antibody (OTI3G9), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-47810
This item is not returnable.
View return policy
Description
IL-6 Monoclonal specifically detects IL-6 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| IL-6 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:50000, Immunohistochemistry 1:50, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:50 | |
| IL6 | |
| Human recombinant protein fragment corresponding to amino acids 29-212 of human IL-6 (NP_000591) produced in E.coli. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| This antibody is specific for Homo sapiens interleukin 6 (interferon, beta 2) (IL6). | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| OTI3G9 | |
| Unconjugated | |
| B-cell differentiation factor, B-cell stimulatory factor 2, BSF2, BSF-2, CDF, CTL differentiation factor, HSF, hybridoma growth factor, IFNB2, IFN-beta-2, IL-6, Interferon beta-2, interleukin 6 (interferon, beta 2), interleukin BSF-2, interleukin-6 | |
| Mouse | |
| 23.5 kDa | |
| 0.1 mL | |
| Adaptive Immunity, Cancer, Cell Cycle and Replication, Cytokine Research, Immunology, Innate Immunity | |
| 3569 | |
| Human, Monkey | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction