missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ IL-6 Antibody (ziltivekimab), Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-28449-100ug
This item is not returnable.
View return policy
Description
IL-6 Monoclonal antibody specifically detects IL-6 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| IL-6 | |
| Monoclonal | |
| Unconjugated | |
| P05231 | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| ziltivekimab | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| B-cell differentiation factor, B-cell stimulatory factor 2, BSF2, BSF-2, CDF, CTL differentiation factor, HSF, hybridoma growth factor, IFNB2, IFN-beta-2, IL-6, Interferon beta-2, interleukin 6 (interferon, beta 2), interleukin BSF-2, interleukin-6 | |
| IL-6/ IFNb2 | |
| 100 μg | |
| Adaptive Immunity, Biologically Active Proteins, Cancer, Cell Cycle and Replication, Cytokine Research, Immunology, Innate Immunity, Neuroscience | |
| 3569 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction