Learn More
CD4 v4 Mouse anti-Human, Brilliant Violet 421, Clone: L120, BD Optibuild™
Mouse Monoclonal Antibody
Brand: BD Biosciences 742650
Description
The L120 monoclonal antibody specifically recognizes CD4. CD4 is an ñ59 kDa type I transmembrane glycoprotein that belongs to the Ig gene superfamily. CD4 is expressed on helper-inducer subset of T lymphocytes and monocytes. The CD4 antigen is involved in the Major Histocompatibility Complex (MHC) class II-restricted recognition of foreign antigens and has been identified as a receptor for HIV. The L120 antibody recognizes amino acids 306-370 of the antigen and does not block the binding of the gp120 of HIV, but inhibits the proliferation of CD4 T lymphocytes to MHC class II antigens. The CD4 antigen is present on most thymocytes, approximately 40% of peripheral blood lymphocytes, and at lower density on monocytes. Peripheral blood CD4+ T lymphocytes are lost over time during HIV infection, and this loss is correlated with disease progression.
The antibody was conjugated to BD Horizon™ BV421 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 407nm and Em Max at 421nm, BD Horizon BV421 can be excited by the violet laser and detected in the standard Pacific Blue™ filter set (eg, 450/50nm filter). BD Horizon BV421 conjugates are very bright, often exhibiting a 10 fold improvement in brightness compared to Pacific Blue conjugates. BV421 is a polymer-based dye excited by the violet laser and is one of the brightest fluorochromes offered by BD Biosciences. Conjugates are typically 10 times brighter than Pacific Blue™ conjugates and are often as bright as or brighter than PE conjugates. Due to nearly identical excitation and emission properties but different spillover characteristics, BD Horizon BV421, Pacific Blue™, and BD Horizon™ V450 cannot be used simultaneously.
Specifications
| CD4 v4 | |
| L120 | |
| Brilliant Violet 421 | |
| L3T4, T-cell surface antigen T4/Leu-3, W3/25, CD4 antigen (p55) | |
| Human Peripheral Blood Lymphocytes | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Monoclonal | |
| 0.2mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Human | |
| IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.