Learn More
IL-6R Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 23457-1-AP-150UL
 View more versions of this product
Product Code. 16812365
  • € 410.00 / 150µL
Description
Interleukin 6 receptor (IL6R), also known as CD126 (Cluster of Differentiation 126), is a type I cytokine receptor. This gene encodes a subunit of the interleukin 6 (IL6) receptor complex. Interleukin 6 is a potent pleiotropic cytokine that regulates cell growth and differentiation and plays an important role in the immune response. The IL6 receptor is a protein complex consisting of this protein and interleukin 6 signal transducer (IL6ST/GP130/IL6-beta), a receptor subunit also shared by many other cytokines. Dysregulated production of IL6 and this receptor are implicated in the pathogenesis of many diseases, such as multiple myeloma, autoimmune diseases and prostate cancer. Alternatively spliced transcript variants encoding distinct isoforms have been reported. A pseudogene of this gene is found on chromosome 9.
Specifications
Specifications
IL-6R
Polyclonal
Unconjugated
IL6R
CD126, gp80, IL 6 receptor subunit alpha, IL 6R 1, IL 6R alpha, IL 6R subunit alpha, IL 6RA, IL-6 R alpha, IL-6R, IL6RA, interleukin 6 receptor, Membrane glycoprotein 80
Rabbit
Antigen Affinity Chromatography
RUO
16194, 24499, 3570
-20°C
Liquid
Immunoprecipitation, Western Blot
0.37 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P08887, P22272, P22273
IL6R, Il6ra
IL-6R Fusion Protein Ag18263
150 μL
Primary
Human, Mouse, Rat