missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-7 Antibody (040), CoraFluor™ 1, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-90485CL1
This item is not returnable.
View return policy
Description
IL-7 Monoclonal antibody specifically detects IL-7 in Mouse samples. It is validated for ELISA
Specifications
| IL-7 | |
| Monoclonal | |
| CoraFluor 1 | |
| IL-7interleukin-7, interleukin 7 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Mouse IL-7 (Accession#: NP_032397.1; Glu26-Ile154). | |
| 0.1 mL | |
| Biologically Active Proteins, Cell Cycle and Replication, Cytokine Research, Hematopoietic Stem Cell Markers, Immunology, Innate Immunity, Stem Cell Markers | |
| 3574 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
| ELISA | |
| 040 | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction