Learn More
IL-7 Antibody (040), PerCP, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90485PCP
This item is not returnable.
View return policy
Description
IL-7 Monoclonal antibody specifically detects IL-7 in Mouse samples. It is validated for ELISA