missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-7 Antibody (MM0410-12T6), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-11724H
This item is not returnable.
View return policy
Description
IL-7 Monoclonal specifically detects IL-7 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| IL-7 | |
| Monoclonal | |
| HRP | |
| PBS with No Preservative | |
| IL7 | |
| Human recombinant IL-7 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| MM0410-12T6 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| IL-7interleukin-7, interleukin 7 | |
| Mouse | |
| Protein G purified | |
| Biologically Active Proteins, Cell Cycle and Replication, Cytokine Research, Hematopoietic Stem Cell Markers, Immunology, Innate Immunity, Stem Cell Markers | |
| 3574 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction