missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IL-7 Monoclonal Antibody (BVD10-40F6)
Rat Monoclonal Antibody
Brand: Invitrogen™ MA183540
This item is not returnable.
View return policy
Description
IL-7 Monoclonal Antibody for ELISA
Interleukin 7 (IL-7) is a hematopoietic cytokine that is constitutively produced by stromal cells from the bone marrow and thymus. IL-7 signals through the IL-7R to promote the differentiation of hematopoietic stem cells to lymphoid precursor cells, which give rise to T cells, B cells, and natural killer cells.
Specifications
| IL-7 | |
| Monoclonal | |
| 0.5 mg/mL | |
| BBS with no preservative | |
| P13232, P16871 | |
| Il7, IL7r | |
| Proprietary Immunogen. | |
| 500 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA | |
| BVD10-40F6 | |
| Unconjugated | |
| Il7 | |
| A630026I06Rik; H-IL-7; hlb368; IL7; Il-7; ILN; Interleukin; interleukin 7; Interleukin7; interleukin-7; M-IL-7 | |
| Rat | |
| Purified | |
| RUO | |
| 3574, 3575 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction