missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IL-7 Monoclonal Antibody (C1), Invitrogen™

Mouse Monoclonal Antibody
€ 185.00 - € 405.00
Specifications
| Antigen | IL-7 |
|---|---|
| Clone | C1 |
| Concentration | 1 mg/mL |
| Applications | Western Blot, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17910003
![]() |
Invitrogen™
MA550927 |
200 μL |
€ 405.00
200µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17979782
|
Invitrogen™
MA550928 |
20 μL |
€ 185.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Interleukin 7 (IL-7) is a hematopoietic cytokine that is constitutively produced by stromal cells from the bone marrow and thymus. IL-7 signals through the IL-7R to promote the differentiation of hematopoietic stem cells to lymphoid precursor cells, which give rise to T cells, B cells, and natural killer cells.Specifications
| IL-7 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4 | |
| A630026I06Rik; H-IL-7; hlb368; IL7; Il-7; ILN; Interleukin; interleukin 7; Interleukin7; interleukin-7; M-IL-7 | |
| Il7, IL7r | |
| IgG1 κ | |
| Protein A/G | |
| Il7 |
| C1 | |
| Western Blot, Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Mouse | |
| Human, Rat | |
| P13232, P16871, P56478 | |
| 25647, 294797, 3574, 3575 | |
| Recombinant Human IL7 protein, Asp26-His177 (Accession #P13232). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title