missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-7 Recombinant Rabbit Monoclonal Antibody (022)

IL-7 Recombinant Rabbit Monoclonal Antibody (22)
Brand: Invitrogen MA530922
This item is not returnable.
View return policy
Description
Specifications
| IL-7 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| P13232, P16871 | |
| Il7, IL7r | |
| Recombinant Human IL7/IL-7/Interleukin-7 Protein (Asp26-His177). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| ELISA | |
| 022 | |
| Unconjugated | |
| Il7 | |
| A630026I06Rik; H-IL-7; hlb368; IL7; Il-7; ILN; Interleukin; interleukin 7; Interleukin7; interleukin-7; M-IL-7 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 3574, 3575 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction