missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-7R alpha/CD127 Antibody (22F3D9), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-22376B
This item is not returnable.
View return policy
Description
IL-7R alpha/CD127 Monoclonal specifically detects IL-7R alpha/CD127 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| IL-7R alpha/CD127 | |
| Monoclonal | |
| Biotin | |
| NP_002176.2 | |
| IL7R | |
| Amino acids 141-237 of human CD127 were used as the immunogen for the antibody. | |
| 0.1 mL | |
| Apoptosis, Cytokine Research, Immunology, Innate Immunity | |
| 3575 | |
| Store at 4C in the dark. | |
| IgG2b λ |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 22F3D9 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| CD127 antigen, CD127ILRA, CDW127, IL-7 receptor subunit alpha, IL-7R subunit alpha, IL7RA, IL-7RA, IL-7R-alpha, interleukin 7 receptor, interleukin 7 receptor alpha chain, interleukin 7 receptor isoform H5-6, interleukin-7 receptor subunit alpha | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction