Learn More
IL-7R alpha/CD127 Antibody (40131), DyLight 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals FAB306K
Product Code. 18589898
  • ONLINE EXCLUSIVE PRICE
    € 724.00
    € 684.60 / 0.10mL
    Save € 39.40
    5% Off
Description
IL-7R alpha/CD127 Monoclonal antibody specifically detects IL-7R alpha/CD127 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IL-7R alpha/CD127
Monoclonal
DyLight 488
50mM Sodium Borate
Mouse
Protein A or G purified from ascites
RUO
Primary
Human
Purified
Western Blot, Flow Cytometry, CyTOF
40131
Western Blot, Flow Cytometry, CyTOF-ready
CD127 antigen, CD127ILRA, CDW127, IL-7 receptor subunit alpha, IL-7R subunit alpha, IL7RA, IL-7RA, IL-7R-alpha, interleukin 7 receptor, interleukin 7 receptor alpha chain, interleukin 7 receptor isoform H5-6, interleukin-7 receptor subunit alpha
Mouse myeloma cell line NS0-derived recombinant human IL-7 R alpha , Extracellular domain
0.1 mL
Apoptosis, Cytokine Research, Immunology, Innate Immunity
3575
Store at 4°C in the dark.