Learn More
Invitrogen™ IL-8 Feline ELISA Kit
Sandwich ELISA Kit
€ 658.00 - € 2640.00
Specifications
Accession Number | Q9XSX5 |
---|---|
Assay Sensitivity | 25 pg/mL |
Assay | Solid-phase sandwich ELISA |
Conjugate | Biotin |
Product Type | ELISA Kit |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15464883
|
Invitrogen™
EFCXCL8 |
96 Tests |
€ 658.00
1 set |
Estimated Shipment: 30-05-2024
Log in to see stock. |
|||||
17104849
|
Invitrogen™
EFCXCL8X5 |
5 x 96 Tests |
€ 2640.00
Each |
Estimated Shipment: 18-06-2024
Log in to see stock. |
|||||
Description
The Feline Interleukin-8/CXCL8 ELISA quantitates Cat IL-8 in feline serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Cat IL-8. Principle of the method The Feline IL-8 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
IL-8 (Interleukin-8) is a 72 amino acid pro-inflammatory CXC chemokine designated CXCL8, secreted by endothelial cells and monocytes, which acts as a major chemoattractant for neutrophils and also T cells and basophils. IL-8, first isolated from osteosarcoma cells, contains the ELR-motif (N-terminal Glu-Leu-Arg amino acid sequence) and signals through the CXCR1 and CXCR2 receptors. Cancer studies have demonstrated a role for IL-8 in the angiogenesis and growth of tumours. Previous nomenclature for IL-8 includes neutrophil activating protein 1 (NAP-1), granulocyte chemotactic protein 1 (GCP-1), monocyte-derived neutrophil-activating peptide (MONAP) and protein 3-10C.Specifications
Q9XSX5 | |
Solid-phase sandwich ELISA | |
ELISA Kit | |
Plasma, Serum, Supernatant | |
96 assays | |
Colorimetric Microplate Reader | |
CXCL8 | |
<10% | |
HRP | |
Extracellular Matrix & Cell Adhesion | |
2°C to 8°C | |
Feline | |
4 hr. 45 min. |
25 pg/mL | |
Biotin | |
GCPR | |
This ELISA pair antibody detects feline IL-8. Other species not determined | |
ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis | |
493836 | |
<12% | |
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
RUO | |
Plasma, 50 μL; Serum, 50 μL; Supernatant, 100 μL | |
IL8 | |
1 hr. 20 min. |