Learn More
IL-9 Antibody (623153), mFluor Violet 500 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals FAB209MFV500
Product Code. 18563786
  • € 607.00 / 0.10mL
Description
IL-9 Monoclonal antibody specifically detects IL-9 in Human samples. It is validated for Flow Cytometry, CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IL-9
Monoclonal
mFluor Violet 500 SE
50mM Sodium Borate
Mouse
0.1 mL
Asthma, Biologically Active Proteins, Cell Cycle and Replication, Cytokine Research, Immunology, Innate Immunity
3578
Store at 4C in the dark.
Flow Cytometry, CyTOF
623153
Intracellular Staining by Flow Cytometry, CyTOF-ready
Cytokine P40, HP40, IL-9homolog of mouse T cell and mast cell growth factor 40, interleukin 9, interleukin-9, P40, p40 cytokine, p40 T-cell and mast cell growth factor, T-cell growth factor P40
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human IL-9, Gln19-Ile144, Accession # P15248
RUO
Primary
Human
IgG2b