missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-9 Antibody (enokizumab), Alexa Fluor™ 750, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals NBP3-28023AF750
This item is not returnable.
View return policy
Description
IL-9 Monoclonal antibody specifically detects IL-9 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| IL-9 | |
| Monoclonal | |
| Alexa Fluor 750 | |
| Cytokine P40, HP40, IL-9homolog of mouse T cell and mast cell growth factor 40, interleukin 9, interleukin-9, P40, p40 cytokine, p40 T-cell and mast cell growth factor, T-cell growth factor P40 | |
| IL-9 | |
| 0.1 mL | |
| Asthma, Biologically Active Proteins, Cell Cycle and Replication, Cytokine Research, Immunology, Innate Immunity | |
| 3578 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| enokizumab | |
| 50mM Sodium Borate | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction